accessibility menu, dialog, popup

UserName

H3CNHH3CCH3H3C

CAS RN 500717-23-7

IUPAC Name: N-(9,9-dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-9H-fluoren-2-amine
Synonyms: N-(9,9-dimethylfluoren-2-yl)-9,9-dimethylfluoren-2-amine
Molecular Weight (g/mol): 401.55
Molecular Formula: C30H27N
InChi Key: LCSMGMWMTSWXDD-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(NC1=CC3=C(C=C1)C1=CC=CC=C1C3(C)C)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results