missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-tert-butylphenyl)iodonium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7119646
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B23801G 1 g
B23805G 5 g
2 options

Catalog No. B23801G

Supplier: TCI America B23801G

Only null left

Chemical Identifiers

CAS 61358-25-6
Molecular Formula C20H26F6IP
Molecular Weight (g/mol) 538.30
MDL Number MFCD06797070
InChI Key CJLLNCQWBHTSCB-UHFFFAOYSA-N
PubChem CID 13906852
IUPAC Name bis(4-tert-butylphenyl)iodanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1

Specifications

Melting Point 175°C
Color White
Quantity 1 g
Formula Weight 538.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-tert-butylphenyl)iodonium Hexafluorophosphate