accessibility menu, dialog, popup

UserName

CAS RN 61358-25-6

CH3H3CCH3ICH3H3CCH3FPFFFFF

CAS RN 61358-25-6

IUPAC Name: bis(4-tert-butylphenyl)iodanium; hexafluoro-λ⁵-phosphanuide
Synonyms: bis(4-tert-butylphenyl)iodanium hexafluorophosphate
Molecular Weight (g/mol): 538.3
Molecular Formula: C20H26F6IP
InChi Key: CJLLNCQWBHTSCB-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results