missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-tert-butylphenyl)amine 90.0+%, TCI America™
$67.11 - $348.57
Chemical Identifiers
| CAS | 4627-22-9 |
|---|---|
| Molecular Formula | C20H27N |
| Molecular Weight (g/mol) | 281.443 |
| InChI Key | OPEKHRGERHDLRK-UHFFFAOYSA-N |
| PubChem CID | 458684 |
| IUPAC Name | 4-tert-butyl-N-(4-tert-butylphenyl)aniline |
| SMILES | CC(C)(C)C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C)(C)C |
Chemical Identifiers
| 4627-22-9 | |
| 281.443 | |
| 458684 | |
| CC(C)(C)C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C)(C)C |
| C20H27N | |
| OPEKHRGERHDLRK-UHFFFAOYSA-N | |
| 4-tert-butyl-N-(4-tert-butylphenyl)aniline |
Specifications
| 4627-22-9 | |
| White-Yellow | |
| C20H27N | |
| 4,4′C-Di-tert-butyldiphenylamine | |
| CC(C)(C)C1=CC=C(C=C1)NC2=CC=C(C=C2)C(C)(C)C | |
| 281.443 | |
| 281.44 | |
| Crystalline Powder |
| 107°C | |
| 195°C | |
| 25 g | |
| OPEKHRGERHDLRK-UHFFFAOYSA-N | |
| 4-tert-butyl-N-(4-tert-butylphenyl)aniline | |
| 458684 | |
| ≥90.0% (GC) | |
| Bis(4-tert-butylphenyl)amine |
Safety and Handling
EINECSNumber : (3)-0135
TSCA : No