accessibility menu, dialog, popup

UserName

CAS RN 4627-22-9

CH3H3CCH3NHH3CH3CCH3

CAS RN 4627-22-9

IUPAC Name: 4-tert-butyl-N-(4-tert-butylphenyl)aniline
Synonyms: 4-tert-butyl-N-(4-tert-butylphenyl)aniline
Molecular Weight (g/mol): 281.44
Molecular Formula: C20H27N
InChi Key: OPEKHRGERHDLRK-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(NC2=CC=C(C=C2)C(C)(C)C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results