missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxybenzyl)amine, 97%, Thermo Scientific Chemicals

$331.88 - $331.88

Chemical Identifiers

CAS 17061-62-0
Molecular Formula C16H19NO2
Molecular Weight (g/mol) 257.333
MDL Number MFCD00277836
InChI Key HBKPDEWGANZHJO-UHFFFAOYSA-N
Synonym bis 4-methoxybenzyl amine, bis-4-methoxy-benzyl-amine, n,n-bis 4-methoxybenzyl amine, bis 4-methoxyphenyl methyl amine, bis-4-methoxybenzyl-amine, benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl, bis-4-methoxybenzyl amine, pubchem23909, di 4-methoxybenzyl amine, maybridge1_007781
PubChem CID 714952
IUPAC Name 1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
SMILES COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5518003
View Documents
Thermo Scientific Chemicals
H5518003
1 g
Each for $331.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17061-62-0
257.333
HBKPDEWGANZHJO-UHFFFAOYSA-N
714952
COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC
C16H19NO2
MFCD00277836
bis 4-methoxybenzyl amine, bis-4-methoxy-benzyl-amine, n,n-bis 4-methoxybenzyl amine, bis 4-methoxyphenyl methyl amine, bis-4-methoxybenzyl-amine, benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl, bis-4-methoxybenzyl amine, pubchem23909, di 4-methoxybenzyl amine, maybridge1_007781
1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine

Specifications

17061-62-0
MFCD00277836
UN3259
HBKPDEWGANZHJO-UHFFFAOYSA-N
1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
714952
97%
C16H19NO2
1 g
bis 4-methoxybenzyl amine, bis-4-methoxy-benzyl-amine, n,n-bis 4-methoxybenzyl amine, bis 4-methoxyphenyl methyl amine, bis-4-methoxybenzyl-amine, benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl, bis-4-methoxybenzyl amine, pubchem23909, di 4-methoxybenzyl amine, maybridge1_007781
COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC
257.333
257.33
Bis(4-methoxybenzyl)amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only