CAS RN 17061-62-0
IUPAC Name:
bis[(4-methoxyphenyl)methyl]amine
Synonyms:
bis[(4-methoxyphenyl)methyl]amine
Molecular Weight (g/mol):
257.33
Molecular Formula:
C16H19NO2
InChi Key:
HBKPDEWGANZHJO-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CNCC2=CC=C(OC)C=C2)C=C1
1
–
1
of
1
results
1
Promotions Available
Bis(4-methoxybenzyl)amine, 97%, Thermo Scientific Chemicals
Bis(4-methoxybenzyl)amine, 97%, Thermo Scientific Chemicals
CAS: 17061-62-0 Molecular Formula: C16H19NO2 Molecular Weight (g/mol): 257.333 MDL Number: MFCD00277836 InChI Key: HBKPDEWGANZHJO-UHFFFAOYSA-N Synonym: bis 4-methoxybenzyl amine,bis-4-methoxy-benzyl-amine,n,n-bis 4-methoxybenzyl amine,bis 4-methoxyphenyl methyl amine,bis-4-methoxybenzyl-amine,benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl,bis-4-methoxybenzyl amine,pubchem23909,di 4-methoxybenzyl amine,maybridge1_007781 PubChem CID: 714952 IUPAC Name: 1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine SMILES: COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC