missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-methacryloylthiophenyl) Sulfide 97.0+%, TCI America™
$829.51 - $2479.19
Chemical Identifiers
| CAS | 129283-82-5 |
|---|---|
| Molecular Formula | C20H18O2S3 |
| Molecular Weight (g/mol) | 386.54 |
| MDL Number | MFCD00142120 |
| InChI Key | SPNAQSNLZHHUIJ-UHFFFAOYSA-N |
Specifications
| 129283-82-5 | |
| White-Yellow | |
| MFCD00142120 | |
| 4,4′C-Thiodibenzenedithiol Dimethacrylate | |
| 386.54 | |
| ≥97.0% (HPLC) | |
| Bis(4-methacryloylthiophenyl) Sulfide |
| 64°C | |
| C20H18O2S3 | |
| 5 g | |
| SPNAQSNLZHHUIJ-UHFFFAOYSA-N | |
| 386.54 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4098
TSCA : No
Recommended Storage : Refrigerator