missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-fluorophenyl) Sulfone 99.0+%, TCI America™
Supplier: TCI America D0537250G
Specifications
| Bis(4-fluorophenyl) Sulfone | |
| 99°C | |
| C12H8F2O2S | |
| 250 g | |
| PLVUIVUKKJTSDM-UHFFFAOYSA-N | |
| 1-fluoro-4-(4-fluorobenzenesulfonyl)benzene | |
| 67842 | |
| ≥99.0% (GC) |
| 383-29-9 | |
| White-Yellow | |
| MFCD00000350 | |
| 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone | |
| FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1 | |
| 254.25 | |
| 254.25 | |
| Crystalline Powder |
Chemical Identifiers
| 383-29-9 | |
| 254.25 | |
| PLVUIVUKKJTSDM-UHFFFAOYSA-N | |
| 67842 | |
| FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1 |
| C12H8F2O2S | |
| MFCD00000350 | |
| 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone | |
| 1-fluoro-4-(4-fluorobenzenesulfonyl)benzene |
Safety and Handling
TSCA : Yes