missing translation for 'onlineSavingsMsg'
Learn More

Bis[4-(dimethylamino)phenyl]methane 98.0+%, TCI America™

Catalog No. B048325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B048325G 25 g
B0483500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B048325G Supplier TCI America Supplier No. B048325G

Chemical Identifiers

CAS 101-61-1
Molecular Formula C17H22N2
Molecular Weight (g/mol) 254.377
MDL Number MFCD00008317
InChI Key JNRLEMMIVRBKJE-UHFFFAOYSA-N
Synonym 4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
PubChem CID 7567
ChEBI CHEBI:34370
IUPAC Name 4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 390°C
Quantity 25 g
Formula Weight 254.38
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[4-(dimethylamino)phenyl]methane
EINECSNumber (3)-0140
RTECSNumber BY5250000
TSCA Yes