accessibility menu, dialog, popup

UserName

H3CNCH3NCH3H3C

CAS RN 101-61-1

IUPAC Name: 4-{[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Synonyms: tetra-base michler ketone, reduced reduced michler's ketone methane base michler's hydride reduced michler ketone michler's ketone, reduced baze michlerova tetrabase michler methane
Molecular Weight (g/mol): 254.38
Molecular Formula: C17H22N2
InChi Key: JNRLEMMIVRBKJE-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(CC2=CC=C(C=C2)N(C)C)C=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)