missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-cyanobenzyl)amine Hydrochloride 98.0+%, TCI America™

Catalog No. B28941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28941G Supplier TCI America Supplier No. B28941G
Only null left

Chemical Identifiers

CAS 1802566-49-9
Molecular Formula C16H14ClN3
Molecular Weight (g/mol) 283.759
InChI Key KUPRRXGOGODEHA-UHFFFAOYSA-N
Synonym alpha,alpha′C-Iminodi-p-tolunitrile Hydrochloride
PubChem CID 44629811
IUPAC Name 4-[[(4-cyanophenyl)methylamino]methyl]benzonitrile;hydrochloride
SMILES C1=CC(=CC=C1CNCC2=CC=C(C=C2)C#N)C#N.Cl

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 283.76
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-cyanobenzyl)amine Hydrochloride