missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chloro-2-nitrophenyl) Disulfide 98.0+%, TCI America™

Catalog No. B42475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42475G 5 g
B424725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42475G Supplier TCI America Supplier No. B42475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2050-66-0
Molecular Formula C12H6Cl2N2O4S2
Molecular Weight (g/mol) 377.21
MDL Number MFCD00047692
InChI Key DESADCWXGJLRSR-UHFFFAOYSA-N
PubChem CID 74907
IUPAC Name 4-chloro-1-[(4-chloro-2-nitrophenyl)disulfanyl]-2-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])SSC2=C(C=C(C=C2)Cl)[N+](=O)[O-]

Specifications

Melting Point 216°C
Quantity 5 g
Synonym 4,4′C-Dichloro-2,2′C-dinitrodiphenyl Disulfide
Formula Weight 377.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-chloro-2-nitrophenyl) Disulfide
EINECSNumber (3)-1131
TSCA Yes