accessibility menu, dialog, popup

UserName

CAS RN 2050-66-0

IUPAC Name: 4-chloro-1-[(4-chloro-2-nitrophenyl)disulfanyl]-2-nitrobenzene
Synonyms: 4-chloro-1-[(4-chloro-2-nitrophenyl)disulfanyl]-2-nitrobenzene
Molecular Weight (g/mol): 377.21
Molecular Formula: C12H6Cl2N2O4S2
InChi Key: DESADCWXGJLRSR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CC=C1SSC1=CC=C(Cl)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results