missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl)amine 98.0+%, TCI America™

Catalog No. B26841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26841G 1 g
B26845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26841G Supplier TCI America Supplier No. B26841G

Chemical Identifiers

CAS 16292-17-4
Molecular Formula C12H9Br2N
Molecular Weight (g/mol) 327.02
MDL Number MFCD00225488
InChI Key VKVHTZNHLOGHGP-UHFFFAOYSA-N
PubChem CID 629950
IUPAC Name 4-bromo-N-(4-bromophenyl)aniline
SMILES BrC1=CC=C(NC2=CC=C(Br)C=C2)C=C1

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 1 g
Synonym 4,4′C-Dibromodiphenylamine
Formula Weight 327.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl)amine
EINECSNumber (3)-0061
TSCA No