accessibility menu, dialog, popup

UserName

BrNHBr

CAS RN 16292-17-4

IUPAC Name: 4-bromo-N-(4-bromophenyl)aniline
Synonyms: 4-bromo-N-(4-bromophenyl)aniline
Molecular Weight (g/mol): 327.02
Molecular Formula: C12H9Br2N
InChi Key: VKVHTZNHLOGHGP-UHFFFAOYSA-N
SMILES: BrC1=CC=C(NC2=CC=C(Br)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results