missing translation for 'onlineSavingsMsg'
Learn More

Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) 98.0+%, TCI America™

$359.76 - $1189.27

Chemical Identifiers

CAS 38951-97-2
Molecular Formula C32H28NiO4S4-4
Molecular Weight (g/mol) 663.505
MDL Number MFCD10567145
InChI Key ADHFORVSZXGTQQ-JZONXAMZSA-J
Synonym Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
PubChem CID 50930260
IUPAC Name (Z)-1,2-bis(4-methoxyphenyl)ethene-1,2-dithiolate;nickel
SMILES COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].[Ni]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B43601G
View Documents
TCI America
B43601G
1 g
Each for $359.76
Only null left
 
B43605G
View Documents
TCI America
B43605G
5 g
Each for $1,189.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

38951-97-2
663.505
ADHFORVSZXGTQQ-JZONXAMZSA-J
50930260
COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].[Ni]
C32H28NiO4S4-4
MFCD10567145
Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
(Z)-1,2-bis(4-methoxyphenyl)ethene-1,2-dithiolate;nickel

Specifications

38951-97-2
C32H28NiO4S4-4
1 g
ADHFORVSZXGTQQ-JZONXAMZSA-J
(Z)-1,2-bis(4-methoxyphenyl)ethene-1,2-dithiolate;nickel
50930260
≥98.0% (HPLC)
Bis[4,4′-dimethoxy(dithiobenzil)]nickel(II)
Black
MFCD10567145
Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].[Ni]
663.505
663.51
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'