accessibility menu, dialog, popup

UserName

NiSSOH3COCH3SSOCH3OCH3

CAS RN 38951-97-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 663.51
Molecular Formula: C32H28NiO4S4
InChi Key: ADHFORVSZXGTQQ-JZONXAMZSA-J
SMILES: [Ni].COC1=CC=C(C=C1)C(\[S-])=C(\[S-])C1=CC=C(OC)C=C1.COC1=CC=C(C=C1)C(\[S-])=C(\[S-])C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)