missing translation for 'onlineSavingsMsg'
Learn More

Bis[4,4'-dimethoxy(dithiobenzil)]nickel(II) 98.0+%, TCI America™

Supplier:  TCI America B43605G

 View more versions of this product

Catalog No. B43605G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis[4,4′-dimethoxy(dithiobenzil)]nickel(II)
Black
MFCD10567145
Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].[Ni]
663.505
663.51
Crystalline Powder
38951-97-2
C32H28NiO4S4-4
5 g
ADHFORVSZXGTQQ-JZONXAMZSA-J
(Z)-1,2-bis(4-methoxyphenyl)ethene-1,2-dithiolate;nickel
50930260
≥98.0% (HPLC)

Chemical Identifiers

38951-97-2
663.505
ADHFORVSZXGTQQ-JZONXAMZSA-J
50930260
COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].COC1=CC=C(C=C1)C(=C(C2=CC=C(C=C2)OC)[S-])[S-].[Ni]
C32H28NiO4S4-4
MFCD10567145
Bis[1,2-bis(4-methoxyphenyl)-1,2-ethenedithiolato]nickel(II)
(Z)-1,2-bis(4-methoxyphenyl)ethene-1,2-dithiolate;nickel
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions