missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane 98.0+%, TCI America™

Catalog No. B1971500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1971500G 500 g
B197125G 25 g
B1971100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1971500G Supplier TCI America Supplier No. B1971500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105391-33-1
Molecular Formula C27H26N2O4
Molecular Weight (g/mol) 442.515
MDL Number MFCD00467754
InChI Key YNSSPVZNXLACMW-UHFFFAOYSA-N
Synonym Methylenebis(3-ethyl-5-methyl-4-maleimidobenzene)
PubChem CID 10950302
IUPAC Name 1-[4-[[4-(2,5-dioxopyrrol-1-yl)-3-ethyl-5-methylphenyl]methyl]-2-ethyl-6-methylphenyl]pyrrole-2,5-dione
SMILES CCC1=CC(=CC(=C1N2C(=O)C=CC2=O)C)CC3=CC(=C(C(=C3)CC)N4C(=O)C=CC4=O)C

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 500 g
Formula Weight 442.52
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-ethyl-5-methyl-4-maleimidophenyl)methane