accessibility menu, dialog, popup

UserName

CAS RN 105391-33-1

IUPAC Name: 1-(4-{[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3-ethyl-5-methylphenyl]methyl}-2-ethyl-6-methylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-{[4-(2,5-dioxopyrrol-1-yl)-3-ethyl-5-methylphenyl]methyl}-2-ethyl-6-methylphenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 442.52
Molecular Formula: C27H26N2O4
InChi Key: YNSSPVZNXLACMW-UHFFFAOYSA-N
SMILES: CCC1=CC(CC2=CC(C)=C(N3C(=O)C=CC3=O)C(CC)=C2)=CC(C)=C1N1C(=O)C=CC1=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results