missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3-aminophenyl) Sulfone 98.0+%, TCI America™
$53.90 - $327.28
Chemical Identifiers
| CAS | 599-61-1 |
|---|---|
| Molecular Formula | C12H12N2O2S |
| Molecular Weight (g/mol) | 248.30 |
| MDL Number | MFCD00007792 |
| InChI Key | LJGHYPLBDBRCRZ-UHFFFAOYSA-N |
| Synonym | 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl |
| PubChem CID | 11741 |
| IUPAC Name | 3-(3-aminobenzenesulfonyl)aniline |
| SMILES | NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1 |
Chemical Identifiers
| 599-61-1 | |
| 248.30 | |
| LJGHYPLBDBRCRZ-UHFFFAOYSA-N | |
| 11741 | |
| NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1 |
| C12H12N2O2S | |
| MFCD00007792 | |
| 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl | |
| 3-(3-aminobenzenesulfonyl)aniline |
Specifications
| 599-61-1 | |
| White-Yellow | |
| MFCD00007792 | |
| 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl | |
| NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1 | |
| 248.30 | |
| 248.30 | |
| Crystalline Powder |
| 172°C | |
| C12H12N2O2S | |
| 25 g | |
| LJGHYPLBDBRCRZ-UHFFFAOYSA-N | |
| 3-(3-aminobenzenesulfonyl)aniline | |
| 11741 | |
| ≥98.0% (T) | |
| Bis(3-aminophenyl) Sulfone |
Safety and Handling
EINECSNumber : (3)-2163
RTECSNumber : BY8800000
TSCA : Yes