missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-aminophenyl) Sulfone 98.0+%, TCI America™

Catalog No. B039425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B039425G 25 g
B0394500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B039425G Supplier TCI America Supplier No. B039425G

Chemical Identifiers

CAS 599-61-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.30
MDL Number MFCD00007792
InChI Key LJGHYPLBDBRCRZ-UHFFFAOYSA-N
Synonym 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl
PubChem CID 11741
IUPAC Name 3-(3-aminobenzenesulfonyl)aniline
SMILES NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 25 g
Formula Weight 248.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-aminophenyl) Sulfone
EINECSNumber (3)-2163
RTECSNumber BY8800000
TSCA Yes