missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-mercaptoethyl) Sulfide 97.0+%, TCI America™

Catalog No. p-7132671
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M133725G 25 g
M1337250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M133725G Supplier TCI America Supplier No. M133725G

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
IUPAC Name 2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
SMILES SCCSCCS

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 g
Formula Weight 154.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-mercaptoethyl) Sulfide
EINECSNumber (2)-3440
TSCA Yes