accessibility menu, dialog, popup

UserName

CAS RN 3570-55-6

IUPAC Name: 2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
Synonyms: bis(2-mercaptoethyl) sulfide
Molecular Weight (g/mol): 154.3
Molecular Formula: C4H10S3
InChi Key: KSJBMDCFYZKAFH-UHFFFAOYSA-N
SMILES: SCCSCCS
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)