missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-mercaptoethyl) Sulfide 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 138°C |
| Quantity | 250 g |
| Formula Weight | 154.30 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(2-mercaptoethyl) Sulfide |
Safety and Handling
| EINECSNumber | (2)-3440 |
| TSCA | Yes |