missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) terephthalate, 97%

Catalog No. p-67996
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402492500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402492500 Supplier Thermo Scientific Chemicals Supplier No. 402492500

CAS: 6422-86-2 | C24H38O4 | 390.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6422-86-2
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.55
MDL Number MFCD00072256
InChI Key RWPICVVBGZBXNA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl terephthalate, kodaflex dotp, di-2-ethylhexyl terephthalate, 1,4-benzenedicarboxylic acid, bis 2-ethylhexyl ester, terephthalic acid, bis 2-ethylhexyl ester, eastman 168, ccris 7052, di 2-ethylhexyl terephthalate, bis 2-ethylhexyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-bis 2-ethylhexyl ester
PubChem CID 22932
IUPAC Name bis(2-ethylhexyl) benzene-1,4-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)C(=O)OCC(CC)CCCC

Specifications

CAS Min % 96
CAS Max % 100.0
Melting Point -67.0°C
Color Colorless
Density 0.9840g/mL
Boiling Point 400.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910
Quantity 250 g
Beilstein 09, IV, 3306
Specific Gravity 0.984
Solubility Information Solubility in water: insoluble
Formula Weight 390.55
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) terephthalate, 97%
Show More Show Less
EINECSNumber 229-176-9
RTECSNumber WZ0883500
TSCA TSCA

RUO – Research Use Only