missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(diphenylphosphino)phenyl] Ether 98.0+%, TCI America™

Catalog No. B286725G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B286725G 25 g
B28675G 5 g
2 options

Catalog No. B286725G

Supplier: TCI America B286725G

Only null left

Chemical Identifiers

CAS 166330-10-5
Molecular Formula C36H28OP2
Molecular Weight (g/mol) 538.57
MDL Number MFCD00233863
InChI Key RYXZOQOZERSHHQ-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinophenyl ether, dpephos, oxydi-2,1-phenylene bis diphenylphosphine, oxybis 2,1-phenylene bis diphenylphosphine, bis 2-diphenylphosphino phenyl ether, 2,2'-bis diphenylphosphino diphenyl ether, 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 4285986
IUPAC Name {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
SMILES O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 25 g
Formula Weight 538.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[2-(diphenylphosphino)phenyl] Ether