accessibility menu, dialog, popup

UserName

POP

CAS RN 166330-10-5

IUPAC Name: {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Synonyms: dpephos
Molecular Weight (g/mol): 538.57
Molecular Formula: C36H28OP2
InChi Key: RYXZOQOZERSHHQ-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)