missing translation for 'onlineSavingsMsg'
Learn More

Bis[(2-diphenylphosphino)phenyl] ether, 98%

Catalog No. p-7047271
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1848103 1 g
AAL1848106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1848103 Supplier Thermo Scientific Chemicals Supplier No. L1848103
Only null left

CAS: 166330-10-5 | C36H28OP2 | 538.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166330-10-5
Molecular Formula C36H28OP2
Molecular Weight (g/mol) 538.57
MDL Number MFCD00233863
InChI Key RYXZOQOZERSHHQ-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinophenyl ether, dpephos, oxydi-2,1-phenylene bis diphenylphosphine, oxybis 2,1-phenylene bis diphenylphosphine, bis 2-diphenylphosphino phenyl ether, 2,2'-bis diphenylphosphino diphenyl ether, 2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
PubChem CID 4285986
IUPAC Name [2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
SMILES O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 184°C to 186°C
Quantity 1 g
Beilstein 7729718
Formula Weight 538.57
Percent Purity 98%
Chemical Name or Material Bis[(2-diphenylphosphino)phenyl] ether
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only