Learn More
Bis[2-(dicyclohexylphosphino)ethyl]amine, 97+%
CAS: 550373-32-5 | C28H53NP2 | 465.69 g/mol
Supplier: Thermo Scientific Chemicals H6026903
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications
| Bis[2-(dicyclohexylphosphino)ethyl]amine | |
| 64°C to 69°C | |
| [(C6H11)2PCH2CH2]2NH | |
| 1 g | |
| Air Sensitive | |
| Insoluble in water. | |
| C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1 | |
| 465.69 | |
| 465.67 |
| 550373-32-5 | |
| C28H53NP2 | |
| MFCD17014019 | |
| UN3259 | |
| bis 2-dicyclohexylphosphino ethyl amine, bis 2-dicyclohexylphosphanyl ethyl amine, 2-dicyclohexylphosphanyl-n-2-dicyclohexylphosphanyl ethyl ethan-1-amine | |
| OQJIHPKBLHGKJN-UHFFFAOYSA-N | |
| 2-dicyclohexylphosphanyl-N-(2-dicyclohexylphosphanylethyl)ethanamine | |
| 58194775 | |
| ≥97% |
Chemical Identifiers
| 550373-32-5 | |
| 465.69 | |
| OQJIHPKBLHGKJN-UHFFFAOYSA-N | |
| 58194775 | |
| C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1 |
| C28H53NP2 | |
| MFCD17014019 | |
| bis 2-dicyclohexylphosphino ethyl amine, bis 2-dicyclohexylphosphanyl ethyl amine, 2-dicyclohexylphosphanyl-n-2-dicyclohexylphosphanyl ethyl ethan-1-amine | |
| 2-dicyclohexylphosphanyl-N-(2-dicyclohexylphosphanylethyl)ethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only