accessibility menu, dialog, popup

UserName

CAS RN 550373-32-5

IUPAC Name: bis[2-(dicyclohexylphosphanyl)ethyl]amine
Synonyms: bis[2-(dicyclohexylphosphanyl)ethyl]amine
Molecular Weight (g/mol): 465.69
Molecular Formula: C28H53NP2
InChi Key: OQJIHPKBLHGKJN-UHFFFAOYSA-N
SMILES: C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)