missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(dicyclohexylphosphino)ethyl]amine, 97+%

Catalog No. p-7040489
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60269MD 250 mg
AAH6026903 1 g
AAH6026906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH60269MD Supplier Thermo Scientific Chemicals Supplier No. H60269MD

CAS: 550373-32-5 | C28H53NP2 | 465.69 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 550373-32-5
Molecular Formula C28H53NP2
Molecular Weight (g/mol) 465.69
MDL Number MFCD17014019
InChI Key OQJIHPKBLHGKJN-UHFFFAOYSA-N
Synonym bis 2-dicyclohexylphosphino ethyl amine, bis 2-dicyclohexylphosphanyl ethyl amine, 2-dicyclohexylphosphanyl-n-2-dicyclohexylphosphanyl ethyl ethan-1-amine
PubChem CID 58194775
IUPAC Name 2-dicyclohexylphosphanyl-N-(2-dicyclohexylphosphanylethyl)ethanamine
SMILES C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 64°C to 69°C
Linear Formula [(C6H11)2PCH2CH2]2NH
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 465.67
Percent Purity ≥97%
Chemical Name or Material Bis[2-(dicyclohexylphosphino)ethyl]amine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only