missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) Ether, 98.5%, Spectrum™ Chemical

$201.91 - $31332.29

Chemical Identifiers

CAS 112-73-2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609075
View Documents
Spectrum Chemical Mfg Cor
B1631100ML
100 mL Amber Glass Bottle
Each for $201.91
Only null left
 
18609078
View Documents
Spectrum Chemical Mfg Cor
B1631500ML
500 mL Amber Glass Bottle
Each for $522.76
Only null left
 
18609076
View Documents
Spectrum Chemical Mfg Cor
B163120LTBL
20 L Steel Can
Each for $4,931.65
Only null left
 
18609077
View Documents
Spectrum Chemical Mfg Cor
B1631200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

112-73-2

Specifications

100 ppm
1
100 mL
Reagent
112-73-2
Amber Glass Bottle
0.985
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.