missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-butoxyethyl) Ether, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B163120LTBL

 View more versions of this product

Catalog No. 18609076


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

100 ppm
1
20 L
Reagent
112-73-2
Steel Can
0.985

Chemical Identifiers

112-73-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.