missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-bromophenyl)acetylene 96.0+%, TCI America™

Catalog No. B49891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49891G 1 g
B49895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B49891G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B49891G

Chemical Identifiers

CAS 38399-13-2
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.03
MDL Number MFCD00238712
InChI Key RVGSIXSCGKMAOK-UHFFFAOYSA-N
PubChem CID 346617
IUPAC Name 1-bromo-2-[2-(2-bromophenyl)ethynyl]benzene
SMILES BrC1=CC=CC=C1C#CC1=CC=CC=C1Br

Specifications

Melting Point 81°C
Color Yellow
Quantity 1 g
Formula Weight 336.03
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-bromophenyl)acetylene