missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-benzamidophenyl) Disulfide 98.0+%, TCI America™

Catalog No. D110225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110225G 25 g
D1102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D110225G Supplier TCI America Supplier No. D110225G

Chemical Identifiers

CAS 135-57-9
Molecular Formula C26H20N2O2S2
Molecular Weight (g/mol) 456.578
MDL Number MFCD00043806
InChI Key ZHMIOPLMFZVSHY-UHFFFAOYSA-N
Synonym bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
PubChem CID 67271
IUPAC Name N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4

Specifications

Melting Point 143°C
Quantity 25 g
Formula Weight 456.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-benzamidophenyl) Disulfide
EINECSNumber (3)-2986
RTECSNumber CV8700000
TSCA Yes