accessibility menu, dialog, popup

UserName

CAS RN 135-57-9

IUPAC Name: N-{2-[(2-benzamidophenyl)disulfanyl]phenyl}benzamide
Synonyms: N-{2-[(2-benzamidophenyl)disulfanyl]phenyl}benzamide
Molecular Weight (g/mol): 456.58
Molecular Formula: C26H20N2O2S2
InChi Key: ZHMIOPLMFZVSHY-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=CC=C1SSC1=CC=CC=C1NC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results