missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,4-dimethylphenyl)amine 98.0+%, TCI America™

Catalog No. p-7123986
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5148200MG 200 mg
B51481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5148200MG Supplier TCI America Supplier No. B5148200MG
Only null left

Chemical Identifiers

CAS 19616-28-5
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.34
MDL Number MFCD16619146
InChI Key MAINCNYZMOMWRA-UHFFFAOYSA-N
Synonym Di-2,4-xylylamine
PubChem CID 21989085
IUPAC Name N-(2,4-dimethylphenyl)-2,4-dimethylaniline
SMILES CC1=CC(C)=C(NC2=C(C)C=C(C)C=C2)C=C1

Specifications

Melting Point 59°C
Color White-Yellow
Quantity 200 mg
Physical Form Crystal-Powder at 20°C
Boiling Point 310°C
Formula Weight 225.34
Percent Purity ≥98.0% (GC)
Chemical Name or Material Bis(2,4-dimethylphenyl)amine