accessibility menu, dialog, popup

UserName

CAS RN 19616-28-5

CH3NHH3CCH3CH3

CAS RN 19616-28-5

IUPAC Name: N-(2,4-dimethylphenyl)-2,4-dimethylaniline
Synonyms: N-(2,4-dimethylphenyl)-2,4-dimethylaniline
Molecular Weight (g/mol): 225.34
Molecular Formula: C16H19N
InChi Key: MAINCNYZMOMWRA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC2=C(C)C=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results