Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
Enables high strength biomagnetic separation
$3086.66 - $4364.02
Specifications
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
BT7102216
View Documents
Agilent BioTek
7102216
96-Well Ring Magnet
Each for $3,086.66
Only null left
BT7102215
View Documents
Agilent BioTek
7102215
384-Well Ring Magnet
Each for $4,364.02
Only null left
Description
Description
  • Magnet ring
  • External
  • For insertion into washer biomagnetic separation carriers
  • Beads pulled to 4-zone ring at well bottom
Specifications
Specifications
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer