Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
Enables high strength biomagnetic separation
Supplier:  Agilent BioTek 7102215
 View more versions of this product
Catalog No. BT7102215
Only null left
Explore available promotions
Description
Description
  • Magnet ring
  • External
  • For insertion into washer biomagnetic separation carriers
  • Beads pulled to 4-zone ring at well bottom
Specifications
Specifications
384-Well Ring Magnet
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer
Product Suggestions
Product Suggestions