missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Automated Microplate Stackers, IVD
GREENER_CHOICE

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. For In Vitro Diagnostic Use.

Supplier:  Agilent BioTek BSTCK2WRSI

 View more versions of this product

Catalog No. BTBSTK2WRSI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity

BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.

Specifications

Chemical Identifiers

18 to 40°C
100/240 V
10 to 85%
LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
11.3 kg
50/60 Hz
CE and TUV marked. RoHS compliant.
One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
<25 lb.

Specifications

50/60 Hz
18 to 40°C
18.5 in.
10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates
One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit
LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers
11.3 kg
18 cm
100/240 V
CE and TUV marked. RoHS compliant.
47 cm
10 to 85%
ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, max. plate height 14.6 mm
<25 lb.
7 in.
BioStack Microplate Stacker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions