missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek BioStack Automated Microplate Stackers, IVD

Description
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity
BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.
Specifications
Specifications
| Hertz | 50/60 Hz |
| Voltage | 100/240 V |
| Operating Temperature Range (Metric) | 18 to 40°C |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Depth (English) | 20.7 in. |
| Depth (Metric) | 52.6 cm |
| Holds | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates |
| Humidity | 10 to 85% |
| Includes | One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit |
| Plate Type | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, max. plate height 14.6 mm |
| Show More |