missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ BIOshell™ lgG, C4 HPLC Column, 1,000 Å, 2.7 μm Particle Size
BIOshell™ IgG 1,000 Å C4 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1, 000 Å) Fused-Core™ particle design.
Supplier: MilliporeSigma™ Supelco™ 63328U
Description
BIOshell™ lgG, C4 columns contain fused-Core particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thin porous shell and the small overall particle size of 2.7-microns. The densely bonded, extensively endcapped dimethylbutyl stationary phase of BIOshell™ IgG 1,000Å C4 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds such as proteins.Specifications
| 4.6 mm | |
| 2.7 μm | |
| 1000 Å | |
| C4 (Butyl) | |
| C4 (Butyl) Phase/Fused-Core Particle Platform | |
| Reversed Phase | |
| Fused-Core HPLC |
| 10 cm | |
| 2 to 9 | |
| 22 m2/g | |
| BIOshell | |
| Yes | |
| Stainless Steel Hardware |
BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.