missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ lgG, C4 Guard Cartridge, 1,000 Å, 2.7 μm Particle Size

Catalog No. 111008438
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111008438 4.6 mm
111008436 2.1 mm
111008437 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111008438 Supplier MilliporeSigma™ Supelco™ Supplier No. 63334U

BIOshell™ IgG 1,000 Å C4 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1, 000 Å) Fused-Core™ particle design.

BIOshell™ lgG, C4 columns contain fused-Core particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5-micron thin porous shell and the small overall particle size of 2.7-microns. The densely bonded, extensively endcapped dimethylbutyl stationary phase of BIOshell™ IgG 1,000Å C4 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds such as proteins.

Specifications

Quantity 3/Pk.
Particle Size 2.7 μm
Length (Metric) 5 mm
Pore Size 1000 Å
Surface Area 22 m2/g
Type Fused-Core HPLC Guard Cartridge
Diameter (Metric) Inner 4.6 mm
Material Stainless Steel Hardware
Phase Reversed Phase
For Use With (Application) HPLC
Endcapped Yes
Product Line BIOshell
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC