missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ IgG C18 HPLC Guard Cartridge, 1,000 Å, 2.7 μm Particle Size

Catalog No. 111008397
Change view
Click to view available options
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
111008397 4.6 mm
111008395 2.1 mm
111008396 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111008397 Supplier MilliporeSigma™ Supelco™ Supplier No. 581361U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BIOshell™ IgG 1,000 Å C18 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1,000 Å) Fused-Core™ particle design.

BIOshell™ IgG 1,000 Å C18 columns contain particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The sterically protected C18 bonded phase of BIOshell™ IgG 1,000 Å C18 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 5 cm
Particle Size 2.7 μm
pH 1 to 8
Pore Size 1000 Å
Quantity 3/Pk.
Surface Area 22 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique UHPLC, HPLC
Product Type Guard Cartridge
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.