Learn More
MilliporeSigma™ Supelco™ BIOshell™ IgG C18 HPLC Column, 1,000 Å, 2.7 μm Particle Size
BIOshell™ IgG 1,000 Å C18 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1,000 Å) Fused-Core™ particle design.
Supplier:  MilliporeSigma™ Supelco™ 582709U
 View more versions of this product
Catalog No. 111007300
Only null left
Explore available promotions
Description
Description
BIOshell™ IgG 1,000 Å C18 columns contain particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The sterically protected C18 bonded phase of BIOshell™ IgG 1,000 Å C18 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.
Specifications
Specifications
4.6 mm
2.7 μm
1000 Å
C18 (octadecyl)
C18 (octadecyl) Phase/Fused-Core Particle Platform
Reversed Phase
3 cm
1 to 8
22 m2/g
BIOshell
Yes
Fused-Core HPLC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.