missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ BIOshell™ IgG C18 HPLC Column, 1,000 Å, 2.7 μm Particle Size

Catalog No. 111006771
Change view
missing translation for 'orderingAttributeHoverText'
Length (Metric):
10 cm
15 cm
25 cm
3 cm
5 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Catalog No. Length (Metric) Diameter (Metric)
111006771 15 cm 2.1 mm
111007297 5 cm 2.1 mm
111006973 10 cm 2.1 mm
111006558 25 cm 2.1 mm
111007298 3 cm 3 mm
111007299 5 cm 3 mm
111006974 10 cm 3 mm
111006772 15 cm 3 mm
111007300 3 cm 4.6 mm
111007301 5 cm 4.6 mm
111006975 10 cm 4.6 mm
111006770 15 cm 4.6 mm
12 options

Catalog No. 111006771

Supplier: MilliporeSigma™ Supelco™ 582703U

Only null left

BIOshell™ IgG 1,000 Å C18 is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1,000 Å) Fused-Core™ particle design.

BIOshell™ IgG 1,000 Å C18 columns contain particles that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The sterically protected C18 bonded phase of BIOshell™ IgG 1,000 Å C18 provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 15 cm
Particle Size 2.7 μm
pH 1 to 8
Pore Size 1000 Å
Surface Area 22 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Fused-Core Particle Platform
Endcapped Yes
Phase Reversed Phase
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.