missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ BIOshell™ A400 Protein C18 Guard Column, 3.4 μm Particle Size
Description
BIOshell™ guard columns are capable of continuous use at pressures up to 9,000 psi (600 bar) with only hand-tightening. Guard cartridges are easily replaced without removing the guard column holder from the flow path. The cartridge is packed with BIOshell™ Fused-Core particles. Order the guard column holder (66841-U) separately.
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 75-35-4 |
| Flash Point | 11°C |
| Linear Formula | C2H2 Cl2 |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Formula Weight | 96.94 |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 μg/mL in methanol |
BIOshell is a trademark of Sigma-Aldrich Co. LLC